2-(Methoxymethoxy)-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxymethoxy-5-(trifluoromethyl)phenylboronic acid

17382-1G 17382-25G 17382-5G
76896-820EA 71.09 USD
76896-820 76896-822 76896-824
2-(Methoxymethoxy)-5-(trifluoromethyl)phenylboronic acid ≥97%
2-(Methoxymethoxy)-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BF₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD11856035
CAS Number: 1256355-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR