2-Fluoro-5-(methoxymethoxy)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-5-(methoxymethoxy)phenyl)boronic acid

17316-1G 17316-25G 17316-500MG 17316-5G
76896-806EA 188.85 USD
76896-806 76896-808 76896-810 76896-812
2-Fluoro-5-(methoxymethoxy)phenylboronic acid ≥97% research grade
2-Fluoro-5-(methoxymethoxy)phenylboronic acid
Formula: C₈H₁₀BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD18837601
CAS Number: 2096332-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR