2-Formyl-6-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17260-1G 17260-25G 17260-500MG 17260-5G
76896-794EA 228.1 USD
76896-794 76896-796 76896-798 76896-800
2-Formyl-6-(trifluoromethyl)phenylboronic acid ≥97%
2-Formyl-6-(trifluoromethyl)phenylboronic acid
Formula: C₈H₆BF₃O₃
MW: 217.938 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295247
CAS Number: 1451392-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR