2,4-Difluoro-3-(phenylmethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

17254-1G 17254-25G 17254-5G 17254-500MG
76896-782EA 112.46 USD
76896-782 76896-784 76896-786 77648-964
2,4-Difluoro-3-(phenylmethoxy)benzaldehyde ≥97%
2,4-Difluoro-3-(phenylmethoxy)benzaldehyde
Formula: C₁₄H₁₀F₂O₂
MW: 248.225 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543645
CAS Number: 947279-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR