3'-Ethoxy-2',4'-difluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Difluoro-3-ethoxy acetophenone

17215-1G 17215-500MG 17215-5G 17215-25G
76896-760EA 725.66 USD
76896-760 76896-762 76896-764 77657-722
3'-Ethoxy-2',4'-difluoroacetophenone ≥97%
3'-Ethoxy-2',4'-difluoroacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258734
CAS Number: 1017778-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR