2,6-Dichloro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

17205-1G 17205-25G 17205-500MG 17205-5G
76896-748EA 132.62 USD
76896-748 76896-750 76896-752 76896-754
2,6-Dichloro-3-methylbenzoic acid ≥97%
2,6-Dichloro-3-methylbenzoic acid
Formula: C₈H₆Cl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00089442
CAS Number: 58671-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR