2-Chloro-4-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17134-1G 17134-25G 17134-5G
76896-736EA 67.9 USD
76896-736 76896-738 76896-740
2-Chloro-4-formylphenylboronic acid ≥97%
2-Chloro-4-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₃
Storage Temperature: Ambient
MDL Number: MFCD18311828
CAS Number: 1063712-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR