2-Fluoro-5-methoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

17119-1G 17119-25G 17119-500MG 17119-5G
76896-720EA 203.7 USD
76896-720 76896-722 76896-724 76896-726
2-Fluoro-5-methoxy-3-methylbenzaldehyde ≥97%
2-Fluoro-5-methoxy-3-methylbenzaldehyde
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
CAS Number: 1056214-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR