2-Fluoro-6-methoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16761-1G 16761-25G 16761-500MG 16761-5G
76896-560EA 290.69 USD
76896-560 76896-562 76896-564 76896-566
2-Fluoro-6-methoxy-3-methylbenzaldehyde ≥97%
2-Fluoro-6-methoxy-3-methylbenzaldehyde
Formula: C₉H₉FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28647478
CAS Number: 1056214-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR