2-Benzyloxy-6-fluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
16726-1G
16726-25G
16726-500MG
16726-5G
76896-538EA
225.98
USD
76896-538
76896-540
76896-542
76896-544
2-Benzyloxy-6-fluorobenzoic acid ≥97%
2-Benzyloxy-6-fluorobenzoic acid
Formula:
C₁₄H₁₁FO₃ MW: 246.234 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520673 CAS Number: 941608-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...