2-Chloro-4-methylpyridine-3-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

16705-1G 16705-500MG 16705-5G
76896-514EA 558.04 USD
76896-514 76896-516 76896-518
2-Chloro-4-methylpyridine-3-boronic acid ≥97% research grade
2-Chloro-4-methylpyridine-3-boronic acid
Formula: C₆H₇BClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07437889
CAS Number: 1029654-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR