2-(Trifluoromethoxy)anisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Methoxy-2-(trifluoromethoxy)benzene, 2-Trifluoromethoxyanisole

16664-25G 16664-5G
76896-496EA 209 USD
76896-496 76896-498
2-(Trifluoromethoxy)anisole ≥97%
2-(Trifluoromethoxy)anisole

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
MW: 192.14 g/mol
Storage Temperature: Ambient
CAS Number: 261952-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR