2,6-Dichloro-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16536-25G 16536-5G
76896-484EA 240.83 USD
76896-484 76896-486
2,6-Dichloro-3-(trifluoromethyl)benzaldehyde ≥97%
2,6-Dichloro-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD11519344
CAS Number: 134741-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR