2-Chloro-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-(trifluoromethyl)-benzaldehyde

16477-25G 16477-5G
76896-466EA 230.22 USD
76896-466 76896-468
2-Chloro-4-(trifluoromethyl)benzaldehyde ≥97%
2-Chloro-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD04112538
CAS Number: 82096-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR