2-Fluoro-6-methyl-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

16430-25G 16430-5G
76896-458EA 418 USD
76896-458 76896-460
2-Fluoro-6-methyl-5-pyridineboronic acid ≥97%
2-Fluoro-6-methyl-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD03411561
CAS Number: 904326-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR