2-Bromo-3-chloro-6-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16418-1G 16418-25G 16418-5G
76896-444EA 96.55 USD
76896-444 76896-446 76896-448
2-Bromo-3-chloro-6-fluorobenzaldehyde ≥97%
2-Bromo-3-chloro-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
MW: 237.454 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520667
CAS Number: 1056264-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR