2-Fluoro-5-methyl-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-methylpyridine-3-boronic acid, (2-Fluoro-5-methylpyridin-3-yl)boronic acid

16404-1G 16404-25G 16404-500MG 16404-5G
76896-428EA 171.87 USD
76896-428 76896-430 76896-432 76896-434
2-Fluoro-5-methyl-3-pyridineboronic acid ≥97%
2-Fluoro-5-methyl-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₂
MW: 154.940 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368857
CAS Number: 1072952-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR