2-Fluoro-(3-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-3-(methoxycarbonyl)phenylboronic acid, (2-Fluoro-3-(methoxycarbonyl)phenyl)boronic acid

16398-1G 16398-25G 16398-5G
76896-418EA 89.12 USD
76896-418 76896-420 76896-422
2-Fluoro-(3-methoxycarbonyl)phenylboronic acid ≥97%
2-Fluoro-(3-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₄
MW: 197.9561 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374985
CAS Number: 1315476-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR