4-Benzyloxy-2-bromo-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1-methyl-4-(phenylmethoxy)benzene

16363-1G 16363-25G 16363-5G
76896-406EA 79.57 USD
76896-406 76896-408 76896-410
4-Benzyloxy-2-bromo-1-methylbenzene ≥97%
4-Benzyloxy-2-bromo-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD09260893
CAS Number: 60710-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR