2,6-Difluoro-N-methoxy-N-methylbenzenecarboxamide ≥97%

Supplier: AOBChem USA

Warning

16341-1G 16341-25G 16341-5G
76896-398EA 84.88 USD
76896-398 76896-400 76896-402
2,6-Difluoro-N-methoxy-N-methylbenzenecarboxamide ≥97%
2,6-Difluoro-N-methoxy-N-methylbenzenecarboxamide
Formula: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD08689758
CAS Number: 937601-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR