2,6-Dichloro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16322-1G 16322-25G 16322-5G
76896-384EA 88.06 USD
76896-384 76896-386 76896-388
2,6-Dichloro-4-methoxybenzaldehyde ≥97%
2,6-Dichloro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12024958
CAS Number: 82772-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR