2-Bromo-6-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16298-25G 16298-5G
76896-376EA 453.01 USD
76896-376 76896-378
2-Bromo-6-fluoro-3-methylbenzaldehyde ≥97%
2-Bromo-6-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD11520666
CAS Number: 154650-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR