2-Bromo-3,6-difluorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
16287-25G
16287-5G
76896-366EA
502.87
USD
76896-366
76896-368
2-Bromo-3,6-difluorobenzaldehyde ≥97%
2-Bromo-3,6-difluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃BrF₂O Storage Temperature: Ambient |
MDL Number:
MFCD12922618 CAS Number: 934987-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...