2,5-Dichloro-3-methoxyphenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

16148-1G 16148-25G 16148-500MG 16148-5G
76896-356EA 290.69 USD
76896-356 76896-358 76896-360 76896-362
2,5-Dichloro-3-methoxyphenylboronic acid ≥95% research grade
2,5-Dichloro-3-methoxyphenylboronic acid
Formula: C₇H₇BCl₂O₃
MW: 220.846 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23379546
CAS Number: 919355-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR