2-Dimethylaminopyrimidine-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

16115-1G 16115-25G 16115-500MG 16115-5G
76896-344EA 155.96 USD
76896-344 76896-346 76896-348 76896-350
2-Dimethylaminopyrimidine-5-carboxylic acid ≥97%
2-Dimethylaminopyrimidine-5-carboxylic acid
Formula: C₇H₉N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06589846
CAS Number: 180283-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR