2,6-Dibromonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dibromopyridine-3-carboxaldehyde

16015-1G 16015-25G 16015-5G
76896-298EA 62.6 USD
76896-298 76896-300 76896-302
2,6-Dibromonicotinaldehyde ≥97%
2,6-Dibromonicotinaldehyde
Formula: C₆H₃Br₂NO
MW: 264.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10574710
CAS Number: 55304-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR