2,6-Dibromo-4-fluorobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

15911-25G 15911-5G
76896-252EA 466.8 USD
76896-252 76896-254
2,6-Dibromo-4-fluorobenzaldehyde ≥97% research grade
2,6-Dibromo-4-fluorobenzaldehyde
Formula: C₇H₃Br₂FO
Storage Temperature: Refrigerator
MDL Number: MFCD13186760
CAS Number: 938467-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR