2-Chloro-4-trifluoromethoxy-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-(trifluoromethoxy)benzaldehyde

15764-1G 15764-25G 15764-500MG 15764-5G
76896-178EA 263.11 USD
76896-178 76896-180 76896-182 76896-184
2-Chloro-4-trifluoromethoxy-benzaldehyde ≥97%
2-Chloro-4-trifluoromethoxy-benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393768
CAS Number: 1079351-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR