2-Bromo-6-chloro-4-(trifluoromethoxy)aniline ≥95%

Supplier: AOBChem USA

Warning

15745-1G 15745-25G 15745-5G
76896-154EA 96.55 USD
76896-154 76896-156 76896-158
2-Bromo-6-chloro-4-(trifluoromethoxy)aniline ≥95%
2-Bromo-6-chloro-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrClF₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD16618925
CAS Number: 1228108-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR