2-Chloro-6-fluoro-3-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

15624-25G 15624-5G
76896-112EA 368.14 USD
76896-112 76896-114
2-Chloro-6-fluoro-3-methoxybenzoic acid ≥95%
2-Chloro-6-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
MW: 204.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115942
CAS Number: 886499-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR