2-Formyl-4-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15555-1G 15555-25G 15555-500MG 15555-5G
76896-070EA 135.8 USD
76896-070 76896-072 76896-074 76896-076
2-Formyl-4-(trifluoromethyl)phenylboronic acid ≥97%
2-Formyl-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₆BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD13195656
CAS Number: 1217501-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR