2-Bromo-5-chloro-4-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-chloropyridine-4-boronic acid, (2-Bromo-5-chloropyridin-4-yl)boronic acid

15502-1G 15502-25G 15502-5G
76896-028EA 88.06 USD
76896-028 76896-030 76896-032
2-Bromo-5-chloro-4-pyridineboronic acid ≥97%
2-Bromo-5-chloro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BBrClNO₂
MW: 236.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064041
CAS Number: 1072952-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR