2-Brom-6-fluoro-3-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15456-1G 15456-25G 15456-5G 15456-500MG
76895-978EA 96.55 USD
76895-978 76895-980 76895-982 77648-750
2-Brom-6-fluoro-3-formylphenylboronic acid ≥97%
2-Brom-6-fluoro-3-formylphenylboronic acid
Formula: C₇H₅BBrFO₃
MW: 246.826 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11856015
CAS Number: 1451392-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR