2-Bromo-1-ethoxy-4-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

15451-1G 15451-25G 15451-500MG 15451-5G
76895-948EA 274.78 USD
76895-948 76895-950 76895-952 76895-954
2-Bromo-1-ethoxy-4-(trifluoromethoxy)benzene ≥97%
2-Bromo-1-ethoxy-4-(trifluoromethoxy)benzene
Formula: C₉H₈BrF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08059525
CAS Number: 200956-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR