(2-Methoxy-5-(trifluoromethoxy)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-5-trifluoromethoxyphenylboronic acid

15445-25G 15445-5G
76895-936EA 537.88 USD
76895-936 76895-938
(2-Methoxy-5-(trifluoromethoxy)phenyl)boronic acid ≥97%
(2-Methoxy-5-(trifluoromethoxy)phenyl)boronic acid
Formula: C₈H₈BF₃O₄
MW: 235.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059517
CAS Number: 290832-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR