2-Bromo-5-chloro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15352-1G 15352-500MG 15352-5G 15352-25G
76895-904EA 495.45 USD
76895-904 76895-906 76895-908 77657-508
2-Bromo-5-chloro-3-methylbenzaldehyde ≥97%
2-Bromo-5-chloro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD34755685
CAS Number: 2092619-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR