2-Chloro-3,5-dibromotoluene ≥97%

Supplier: AOBChem USA

Warning

15346-1G 15346-25G 15346-500MG 15346-5G
76895-894EA 126.25 USD
76895-894 76895-896 76895-898 76895-900
2-Chloro-3,5-dibromotoluene ≥97%
2-Chloro-3,5-dibromotoluene
Formula: C₇H₅Br₂Cl
MW: 284.376 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878154
CAS Number: 874513-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR