5-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Formyl-4-methoxyphenylboronic acid pinacol ester
15322-25G
15322-5G
76895-868EA
421.18
USD
76895-868
76895-870
5-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥95%
5-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formula:
C₁₄H₁₉BO₄ MW: 262.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11044929 CAS Number: 1132669-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...