2-Chloro-3-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15262-25G 15262-5G
76895-840EA 524.09 USD
76895-840 76895-842
2-Chloro-3-(trifluoromethyl)phenylboronic acid ≥97%
2-Chloro-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.3726 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258745
CAS Number: 957061-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR