2-Methyl-4-(trifluoromethyl)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

15248-1G 15248-25G 15248-5G
76895-832EA 94.43 USD
76895-832 76895-834 76895-836
2-Methyl-4-(trifluoromethyl)benzaldehyde ≥97% research grade
2-Methyl-4-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O
MW: 188.146 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18399853
CAS Number: 888739-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR