2,6-Dichloro-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dichloro-4-trifluoromethylbenzaldehyde

15242-1G 15242-25G 15242-5G
76895-820EA 75.33 USD
76895-820 76895-822 76895-824
2,6-Dichloro-4-(trifluoromethyl)benzaldehyde ≥97%
2,6-Dichloro-4-(trifluoromethyl)benzaldehyde
Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20441469
CAS Number: 118754-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR