2-Chloro-4,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15237-25G 15237-5G
76895-812EA 386.17 USD
76895-812 76895-814
2-Chloro-4,5-difluorobenzaldehyde ≥97%
2-Chloro-4,5-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MW: 176.5500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782050
CAS Number: 165047-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR