2-Bromo-4-chloro-6-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15195-1G 15195-25G 15195-5G
76895-776EA 96.55 USD
76895-776 76895-778 76895-780
2-Bromo-4-chloro-6-fluorobenzaldehyde ≥95%
2-Bromo-4-chloro-6-fluorobenzaldehyde
Formula: C₇H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD12400895
CAS Number: 1135531-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR