2-Chloro-6-fluoro-3-formylphenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
15191-1G
15191-25G
15191-5G
15191-500MG
76895-762EA
96.55
USD
76895-762
76895-764
76895-766
77644-660
2-Chloro-6-fluoro-3-formylphenylboronic acid ≥97%
2-Chloro-6-fluoro-3-formylphenylboronic acid
Formula:
C₇H₅BClFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD11856014 CAS Number: 1451392-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...