2-Bromo-3-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

15184-1G 15184-25G 15184-500MG 15184-5G
76895-748EA 150.65 USD
76895-748 76895-750 76895-752 76895-754
2-Bromo-3-(trifluoromethyl)benzaldehyde ≥95%
2-Bromo-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
MW: 253.016 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520657
CAS Number: 1114808-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR