2,6-Dichloro-4-methylaniline ≥97%

Supplier: AOBChem USA

Warning

15177-1G 15177-25G 15177-5G
76895-720EA 64.72 USD
76895-720 76895-722 76895-724
2,6-Dichloro-4-methylaniline ≥97%
2,6-Dichloro-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD00190134
CAS Number: 56461-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR