2-Bromo-3-butoxy-6-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-3-butoxy-6-fluorophenyl)boronic acid

15160-1G 15160-25G 15160-5G
76895-712EA 64.72 USD
76895-712 76895-714 76895-716
2-Bromo-3-butoxy-6-fluorophenylboronic acid ≥97%
2-Bromo-3-butoxy-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BBrFO₃
MW: 290.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08276824
CAS Number: 1072951-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR