2-Bromo-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-5-(trifluoromethyl)phenyl)boronic acid, 2-Bromo-5-trifluoromethylphenylboronic acid

15158-25G 15158-5G
76895-704EA 559.1 USD
76895-704 76895-706
2-Bromo-5-(trifluoromethyl)phenylboronic acid ≥97%
2-Bromo-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrF₃O₂
MW: 268.82 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258742
CAS Number: 957034-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR