2-Bromo-6-chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-6-chlorophenyl)boronic acid

15156-25G 15156-5G
76895-698EA 488.02 USD
76895-698 76895-700
2-Bromo-6-chlorophenylboronic acid ≥97%
2-Bromo-6-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrClO₂
MW: 235.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701777
CAS Number: 1107580-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR