2-Bromo-6-fluoro-3-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-6-fluoro-3-methylphenyl)boronic acid

15142-25G 15142-5G
76895-680EA 204.76 USD
76895-680 76895-682
2-Bromo-6-fluoro-3-methylphenylboronic acid ≥97%
2-Bromo-6-fluoro-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₂
Storage Temperature: Ambient
MDL Number: MFCD09800874
CAS Number: 957121-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR